Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIB3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | CIB3 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CIB3 Polyclonal specifically detects CIB3 in Human samples. It is validated for Western Blot.Specifications
CIB3 | |
Polyclonal | |
Rabbit | |
Q96Q77 | |
117286 | |
Synthetic peptides corresponding to CIB3(calcium and integrin binding family member 3) The peptide sequence was selected from the N terminal of CIB3. Peptide sequence QDLAPQLVPLDYTTCPDVKVPYELIGSMPELKDNPFRQRIAQVFSEDGDG. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
calcium and integrin binding family member 3, calcium and integrin-binding family member 3, DNA-dependent protein kinase catalytic subunit-interacting protein 3, Kinase-interacting protein 3, KIP 3, KIP3MGC96922, MGC138405, MGC142151 | |
CIB3 | |
IgG | |
22 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title