Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ciclopirox Olamine, USP, 97.5-101.5%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
41621-49-2
C14H24N2O3
268.36
MBRHNTMUYWQHMR-UHFFFAOYSA-N
2-aminoethan-1-ol; 6-cyclohexyl-1-hydroxy-4-methyl-1,2-dihydropyridin-2-one
NCCO.CC1=CC(=O)N(O)C(=C1)C1CCCCC1
41621-49-2
C14H24N2O3
268.36
MBRHNTMUYWQHMR-UHFFFAOYSA-N
2-aminoethan-1-ol; 6-cyclohexyl-1-hydroxy-4-methyl-1,2-dihydropyridin-2-one
NCCO.CC1=CC(=O)N(O)C(=C1)C1CCCCC1

C1486, 41621-49-2, C12H17NO2•C2H7NO

Spectrum™ Ciclopirox Olamine, USP is a synthetic antifungal agent for topical dermatologic treatment of superficial mycoses. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.1%
Quantity 25 g
Packaging Amber Glass Bottle
CAS Max % 100%
Percent Purity 97.5-101.5%