Learn More
Tocris Bioscience CID 16020046
Selective GPR55 antagonist
Manufacturer: Tocris Bioscience 4959/10
This item is not returnable.
View return policy
Description
Selective GPR55 antagonist. Inhibits LPI-induced Ca2+ signaling (IC50 = 0.21µM in HEK-GPR55 cells), ERK1/2 phosphorylation and GPR55-mediated transcription factor activation. Displays weak inhibition of acetylcholinesterase,µ-opioid receptor, KCNH2 and hERG. Decreases LPI-induced GPR55 internalization. Reduces experimental intestinal inflammation in mice.Specifications
CID 16020046 | |
>99 | |
10mg |
CID 16020046 | |
425.44 |