Tocris™ CID 16020046

Selective GPR55 antagonist

Manufacturer: Tocris™ 4959/50

 View more versions of this product

Catalog No. 49-595-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors CID 16020046
    Purity >99
    Molecular Weight 425.44
    Quantity 50mg
    Product Type CID 16020046

    Selective GPR55 antagonist. Inhibits LPI-induced Ca2+ signaling (IC50 = 0.21µM in HEK-GPR55 cells), ERK1/2 phosphorylation and GPR55-mediated transcription factor activation. Displays weak inhibition of acetylcholinesterase,µ-opioid receptor, KCNH2 and hERG. Decreases LPI-induced GPR55 internalization. Reduces experimental intestinal inflammation in mice.

    • Chemical Name: 4-[4,6-Dihydro-4-(3-hydroxyphenyl)-3-(4-methylphenyl)-6-oxopyrrolo[3,4-c]pyrazol-5(1H)-yl]benzoic acid
    • Formula: C25H19N3O4
    • CAS Number: 834903-43-4
    • Solubility: Soluble to 100mM in DMSO
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit