Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CID 2011756, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
638156-11-3
C22H21ClN2O3
396.871
XQJWTJLJEYIUDZ-UHFFFAOYSA-N
cid,5-3-chlorophenyl-n-4-morpholinomethyl phenyl furan-2-carboxamide,5-3-chlorophenyl-n-4-morpholin-4-ylmethyl phenyl furan-2-carboxamide,5-3-chlorophenyl-n-4-4-morpholinylmethyl phenyl-2-furancarboxamide,5-3-chlorophenyl-n-4-morpholinomethyl-phenyl furan-2-carboxamide,5-3-chloro-phenyl-furan-2-carboxylic acid 4-morpholin-4-ylmethyl-phenyl-amide,qcr-12,cid hplc,5-3-chlorophenyl-n-4-morpholinomethyl phenyl-2-furamide,5-3-chlorophenyl-n-4-morpholin-4-ymethyl phenyl furan-2-carboxamide
2011756
5-(3-chlorophenyl)-N-[4-(morpholin-4-ylmethyl)phenyl]furan-2-carboxamide
C1COCCN1CC2=CC=C(C=C2)NC(=O)C3=CC=C(O3)C4=CC(=CC=C4)Cl
This item is not returnable. View return policy
638156-11-3
C22H21ClN2O3
396.871
XQJWTJLJEYIUDZ-UHFFFAOYSA-N
cid,5-3-chlorophenyl-n-4-morpholinomethyl phenyl furan-2-carboxamide,5-3-chlorophenyl-n-4-morpholin-4-ylmethyl phenyl furan-2-carboxamide,5-3-chlorophenyl-n-4-4-morpholinylmethyl phenyl-2-furancarboxamide,5-3-chlorophenyl-n-4-morpholinomethyl-phenyl furan-2-carboxamide,5-3-chloro-phenyl-furan-2-carboxylic acid 4-morpholin-4-ylmethyl-phenyl-amide,qcr-12,cid hplc,5-3-chlorophenyl-n-4-morpholinomethyl phenyl-2-furamide,5-3-chlorophenyl-n-4-morpholin-4-ymethyl phenyl furan-2-carboxamide
2011756
5-(3-chlorophenyl)-N-[4-(morpholin-4-ylmethyl)phenyl]furan-2-carboxamide
C1COCCN1CC2=CC=C(C=C2)NC(=O)C3=CC=C(O3)C4=CC(=CC=C4)Cl

Pan protein kinase D (PKD) inhibitor; cell permeable

Protein kinase D (PKD) inhibitor (IC50 values are 0.6, 0.7 and 3.2 μM for PKD2, PKD3 and PKD1 respectively). Cell permeable (EC50 = 10 μM for PKD1 inhibition). ATP-competitive.
Quantity 10mg
Formula Weight 396.87
Percent Purity >99%
Chemical Name or Material CID 2011756

Recommended Storage : Store at Room Temperature