Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIDEC Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100430H
Description
CIDEC Polyclonal antibody specifically detects CIDEC in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
CIDEC | |
Polyclonal | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
HRP | |
cell death activator CIDE-3, cell death-inducing DFFA-like effector c, Cell death-inducing DFFA-like effector protein C, CIDE-3, CIDEX, Fat-specific protein FSP27 homolog, FLJ20871, Fsp27, FSP27fat specific protein 27 | |
Residues 10-25 LLYPKSLSRHVSVRTS] of the 27 kDa human FSP27 (CIDE-3) protein. | |
0.1 mL | |
Apoptosis, Cancer, DNA Repair | |
63924 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction