Learn More
CIITA Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259074
Catalog No. NBP259074
Add to cart
Description
Description
CIITA Polyclonal antibody specifically detects CIITA in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
CIITA
0.95 mg/ml
Western Blot 1:500-1:3000, ELISA 1:5000-1:25000
C2TA, C2TAMHC class II transactivator type III, CIITAIV, class II, major histocompatibility complex, transactivator, MHC class II transactivator, MHC2TANLR family, acid domain containing, NLRA, nucleotide-binding oligomerization domain, leucine rich repeat and acid domaincontaining
Rabbit
IgG
0.1 mg
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Detects human CIITA
ELISA, Western Blot
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
CIITA
Anti-CIITA was produced by repeated immunizations with CIITA peptide corresponding to a region near the N-terminus of the human protein conjugated to Keyhole Limpet Hemocyanin (KLH).
Ion exchange chromatography
RUO
Primary
4261
Human