Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cilastatin sodium, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Dipeptidase inhibitor (LTDase, leukotriene D4 hydrolase, dehydropeptidase I) that displays a Ki value of 0.11 μM. Inhibits metabolism of LTD4 to LTE4 and the hydrolysis of β-lactam antibiotics. Nephroprotective; reduces toxic accumulation of cyclosporin A in kidney proximal tubule epithelial cells.
Specifications
Quantity | 50 mg |
Formula Weight | 380.43 |
Percent Purity | >99% |
Chemical Name or Material | Cilastatin sodium |
Safety and Handling
Recommended Storage : Desiccate at -20°C