Learn More
Abcam Cilnidipine, Dual L- and N-type Ca2+ channel blocker, 10MG
Supplier:  Abcam AB14103810MG
Encompass_Preferred
MW 492.53 g/mol, Purity >98%. Dual L- and N-type Ca²+ channel blocker (IC₅₀ values are 100 and 200 nM, respectively). Shows antihypertensive, antinociceptive and neuroprotective effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6956