Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cilnidipine, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Dual L- and N-type calcium channel blocker that displays antihypertensive, sympatholytic and neuroprotective activity. Lowers mean blood pressure and reduces the size of cerebral infarction in the rat model of focal brain ischemia.
Specifications
Quantity | 50 mg |
Formula Weight | 492.52 |
Percent Purity | >99% |
Chemical Name or Material | Cilnidipine |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction