Learn More
Selleck Chemical LLCSupplier Diversity Partner Cilomilast 10mg 153259-65-5 SB-207499  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S145510MG
Cilomilast (SB-207499) is a potent PDE4 inhibitor with IC50 of about 110 nM, has anti-inflammatory activity and low central nervous system activity. Phase 3.
Catalog No. 50-633-9