Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cilostamide, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

68550-75-4
C20H26N2O3
342.439
UIAYVIIHMORPSJ-UHFFFAOYSA-N
N-Cyclohexyl-N-methyl-4-(1, 2-dihydro-2-oxo-6-quinolyloxy)butyramide
2753

PDE3 inhibitor

Selective inhibitor of type III phosphodiesterase (PDE3). Displays moderate selectivity for PDE3A isozyme vs. PDE3B (IC50 values are 0.027 and 0.050 μM for PDE3A and PDE3B respectively). Inhibits ADP-induced platelet aggregation (IC50 = 16.8 μM); antithrombotic. Also available as part of the Phosphodiesterase Inhibitor Tocriset™ .

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
SMILES CN(C1CCCCC1)C(=O)CCCOC2=CC3=C(C=C2)NC(=O)C=C3
IUPAC Name N-cyclohexyl-N-methyl-4-[(2-oxo-1H-quinolin-6-yl)oxy]butanamide
Formula Weight 342.44
Chemical Name or Material Cilostamide
Grade >98%

Recommended Storage : Store at Room Temperature