Learn More
Thermo Scientific™ Cimarec+™ Stirring Hotplate Series, Ceramic PROMO
Get precise stirring control, exceptional safety and temperature performance for your routine protocols.
Manufacturer:  Thermo Scientific™ SP8885710012N
 View more versions of this product
Catalog No. SP88857100F
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Advanced technology
  • Stir-Trac features innovative engineering for slow-speed stirring, consistent speed control, and strong magnetic coupling
  • Temperature adjustable in 1 degree increments
  • Hot Top warning system protects from accidental burns with prominent display when heating surface is above 50°C (122°F)
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Easy-to-read LED display for heating and stirring
  • Raised display design protects electronics from spills
  • Optional temperature probe and splash guard protection shield
  • Variable sizes for different sample volumes
Warranty: 3 years
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.