Learn More
Thermo Scientific™ Cimarec+™ Stirring Hotplates Series, Ceramic, 10.25 in. Products for Science Education
Digital stirring hotplates offer precise stirring control, exceptional safety and temperature performance for your routine protocols.
Manufacturer:  Thermo Scientific™ SP88850100/EMD
Catalog No. S23627
  Check Availability
Add to cart
Disclaimers:
Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Advanced technology
  • Stir-Trac features innovative engineering for slow-speed stirring, consistent speed control, and strong magnetic coupling
  • Temperature adjustable in 1 degree increments
  • Hot Top warning system protects from accidental burns with prominent display when heating surface is above 50°C (122°F)
Flexible, robust design
  • Ceramic top plate
  • Easy-to-read LED display for heating and stirring
  • Raised display design protects electronics from spills
  • Optional temperature probe and splash guard protection shield
Description & Specifications
Specifications
50 to 1,500rpm
10.25 x 10.25 in
Ceramic
1 power cord
7.5kg
16.2 x 11.3 x 4 in. (41.1 x 28.7 x 10.2cm)
400°C (Max Temperature)
26.0 x 26.0cm
cCSAus, CE
16.5 lb
50 to 1,500rpm
100-120V; 50/60Hz