Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cimetidine Hydrochloride, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum

C1673, 70059-30-2, C10H16N6S•HCl

Spectrum™ Cimetidine Hydrochloride, USP
Packaging Fiber Drum
Loss on Drying 0.005
Ignition Residue 0.002
Quantity 25 kg
CAS 70059-30-2
CAS Max % 1
Molecular Formula C10H17ClN6S
InChI Key QJHCNBWLPSXHBL-UHFFFAOYSA-N
SMILES Cl.CN=C(NCCSCC1=C(C)NC=N1)NC#N
IUPAC Name N-cyano-N''-methyl-N'-(2-{[(5-methyl-1H-imidazol-4-yl)methyl]sulfanyl}ethyl)guanidine hydrochloride
Molecular Weight (g/mol) 288.80
Percent Purity 98 to 102%
Show More Show Less