Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cimetidine, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
51481-61-9
C10H16N6S
252.34
AQIXAKUUQRKLND-UHFFFAOYSA-N
N-cyano-N''-methyl-N'-(2-{[(5-methyl-1H-imidazol-4-yl)methyl]sulfanyl}ethyl)guanidine
CN=C(NCCSCC1=C(C)NC=N1)NC#N
51481-61-9
C10H16N6S
252.34
AQIXAKUUQRKLND-UHFFFAOYSA-N
N-cyano-N''-methyl-N'-(2-{[(5-methyl-1H-imidazol-4-yl)methyl]sulfanyl}ethyl)guanidine
CN=C(NCCSCC1=C(C)NC=N1)NC#N

C1491, 51481-61-9, C10H16N6S

Spectrum™ Cimetidine, USP is a histamine and H2 receptor antagonist used to treat peptic ulcers and heartburn. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Quantity 25 kg
Melting Range 139-144°C
Ignition Residue 0.2%
Loss on Drying 1%
Packaging Fiber Drum
CAS Max % 100%
Percent Purity 98-102%
Grade USP