Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Cinacalcet hydrochloride

Low molecular weight, organic compound that may help regulate biological process.

$93.70 - $393.00

Specifications

CAS 364782-34-3
Formula Weight 393.87g/mol
For Use With (Application) Calcium-Sensing Receptor Agonists
Molecular Formula C22H22F3N.HCl
Storage Requirements Store at −20°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
61-701-0 View Documents Tocris Bioscience™
6170/10
10mg Each for $93.70
Add to cart
 
61-705-0 View Documents Tocris Bioscience™
6170/50
50mg Each for $393.00
Add to cart
 
Description

Description

Calcium-sensing receptor (CaSR) allosteric agonist. Activates CaSR signaling in cellular proliferation and phosphatidylinositol (PI) hydrolysis assays. Reduces serum parathyroid hormone levels in rats. Also potent CYP2D2 inhibitor (IC50 = 87 nM) and L-type calcium channel blocker. Orally bioavailable.
Specifications

Specifications

364782-34-3
Calcium-Sensing Receptor Agonists
Store at −20°C
393.87g/mol
C22H22F3N.HCl
Calcium-Sensing Receptor

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit