Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cinanserin hydrochloride, 99+%

Click to view available options
Quantity:
10mg
50mg
1166-34-3
C20H25ClN2OS
376.943
MFCD00055609
LXGJPDKYMJJWRB-IERUDJENSA-N
6433141
(E)-N-[2-[3-(dimethylamino)propylsulfanyl]phenyl]-3-phenylprop-2-enamide;hydrochloride
CN(C)CCCSC1=CC=CC=C1NC(=O)C=CC2=CC=CC=C2.Cl
1166-34-3
C20H25ClN2OS
376.943
MFCD00055609
LXGJPDKYMJJWRB-IERUDJENSA-N
6433141
(E)-N-[2-[3-(dimethylamino)propylsulfanyl]phenyl]-3-phenylprop-2-enamide;hydrochloride
CN(C)CCCSC1=CC=CC=C1NC(=O)C=CC2=CC=CC=C2.Cl

A 5-HT antagonist

Quantity 10mg
Color White
Solubility Information Soluble to 50 mM in water
Formula Weight 376.94
Physical Form Solid
Percent Purity ≥99%
Chemical Name or Material Cinanserin hydrochloride

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

RTECSNumber : UC6323000

TSCA : No

Recommended Storage : Ambient temperatures