Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (-)-Cinchonidine, 99% (total base), may cont. up to 5% quinine

Click to view available options
Quantity:
25g
100g
485-71-2
C19H22N2O
294.398
MFCD00006783
KMPWYEUPVWOPIM-HVIVRAMPSA-N
cinchonidine
45358337
[(2R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-quinolin-4-ylmethanol
C=CC1CN2CCC1CC2C(C3=CC=NC4=CC=CC=C34)O
485-71-2
C19H22N2O
294.398
MFCD00006783
KMPWYEUPVWOPIM-HVIVRAMPSA-N
cinchonidine
45358337
[(2R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-quinolin-4-ylmethanol
C=CC1CN2CCC1CC2C(C3=CC=NC4=CC=CC=C34)O
Applications
It finds its uses as an antimalarial compound. It is a lysosomotropic agent, inhibits Plasmodium falciparum, interferes with ferriprotoporphyrin formation and is a weak SERT inhibitor.

Solubility
Insoluble in water. Soluble in alcohol and chloroform.

Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Sensitivity Light sensitive
Quantity 100g
Optical Rotation −109° (c=0.5 in Ethanol)
Melting Point 201°C to 206°C
Beilstein 89690
Merck Index 14,2286
Solubility Information Insoluble in water. Soluble in alcohol and chloroform.
Formula Weight 294.39
Percent Purity 99%
Chemical Name or Material (-)-Cinchonidine
Assay (total base), may cont. up to 5% quinine
Show More Show Less

GHS H Statement
H334-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause an allergic skin reaction.

P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c

H302-H317

EINECSNumber : 207-622-3

RTECSNumber : GD3500000

TSCA : Yes

Recommended Storage : Ambient temperatures