Learn More
Cincinnati Surgical Non-Sterile Carbon Blades
Packaged with VCI Paper to protect blade from corrosion
Manufacturer:  Cincinnati Surgical 0260
 View more versions of this product
Catalog No. 19-200-259
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Made from high-grade British surgical carbon steel
  • Narrow grinding angle resulting in increased initial keenness
  • Medium surface finish
  • 200% visual inspection
  • Strong foil package
Specifications
Specifications
60
Non-sterile
British carbon steel
Medium surface finish