Learn More
Cincinnati Surgical Sterile Stainless Steel Blades and Scalpels
Packaged with VCI Paper to protect the blade from corrosion
Manufacturer: Cincinnati Surgical 0520
Description
- Made from British stainless steel
- 200% visual inspection
- Narrow grinding angle resulting in increased initial keenness
- Medium surface finish
- Strong foil package
- Handles are made of plastic
Specifications
#20 | |
British stainless steel | |
Flexible blade cover |
Sterile | |
Medium surface finish | |
10/Pk. |