Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cinnamon Bark Oil, Spectrum™

Click to view available options
Quantity:
25 mL
100 mL
25 lb.
Packaging:
Amber Glass Bottle
Steel Pail
8015-91-6
8015-91-6
Quantity 25 lb.
CAS Max % 1
Packaging Steel Pail
Grade Reagent

CAUTION: For manufacturing, processing or repacking. Not for food use. Read and understand the label and Safety Data Sheet (SDS) prior to use.