Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cinnamonitrile, 97%, predominantly trans

Click to view available options
Quantity:
25g
100g
500g
1885-38-7
C9H7N
129.162
MFCD00001930
ZWKNLRXFUTWSOY-QPJJXVBHSA-N
cinnamonitrile,trans-cinnamonitrile,styryl cyanide,cinnamyl nitrile,3-phenylacrylonitrile,beta-cyanostyrene,cinnamonitrile, e,3-phenyl-2-propenenitrile,acrylonitrile, 3-phenyl,e-3-phenylprop-2-enenitrile
1550846
(E)-3-phenylprop-2-enenitrile
C1=CC=C(C=C1)C=CC#N
1885-38-7
C9H7N
129.162
MFCD00001930
ZWKNLRXFUTWSOY-QPJJXVBHSA-N
cinnamonitrile,trans-cinnamonitrile,styryl cyanide,cinnamyl nitrile,3-phenylacrylonitrile,beta-cyanostyrene,cinnamonitrile, e,3-phenyl-2-propenenitrile,acrylonitrile, 3-phenyl,e-3-phenylprop-2-enenitrile
1550846
(E)-3-phenylprop-2-enenitrile
C1=CC=C(C=C1)C=CC#N
Applications
Cinnamonitrile is a parent compound whose derivatives can be applied as corrosion inhibitors for API J55 steel.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Air & light sensitive. Store in dark. Incompatible with oxidizing agents.
Density 1.029
Sensitivity Air and light sensitive
Refractive Index 1.602
Quantity 500g
Boiling Point 253°C to 254°C
Flash Point 126°C (259°F)
Melting Point 18°C to 20°C
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 129.16
Percent Purity 97%
Chemical Name or Material Cinnamonitrile, predominantly trans
Show More Show Less

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 217-552-5

RTECSNumber : UD1440000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen