Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cinnamyl chloride, 95% trans, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
2687-12-9
C9H9Cl
152.62
MFCD00000986
IWTYTFSSTWXZFU-QPJJXVBHSA-N
cinnamyl chloride,3-chloroprop-1-en-1-yl benzene,3-chloro-1-phenyl-1-propene,trans-cinnamyl chloride,1e-3-chloroprop-1-en-1-yl benzene,benzene, 3-chloropropenyl,e-3-chloroprop-1-enyl benzene,unii-2i1adl56tx,2i1adl56tx,cinnamylchloride
639658
[(E)-3-chloroprop-1-enyl]benzene
C1=CC=C(C=C1)C=CCCl
2687-12-9
C9H9Cl
152.62
MFCD00000986
IWTYTFSSTWXZFU-QPJJXVBHSA-N
cinnamyl chloride,3-chloroprop-1-en-1-yl benzene,3-chloro-1-phenyl-1-propene,trans-cinnamyl chloride,1e-3-chloroprop-1-en-1-yl benzene,benzene, 3-chloropropenyl,e-3-chloroprop-1-enyl benzene,unii-2i1adl56tx,2i1adl56tx,cinnamylchloride
639658
[(E)-3-chloroprop-1-enyl]benzene
C1=CC=C(C=C1)C=CCCl
Flash Point 79°C
Specific Gravity 1.096
Water 0.1% max. (K.F.)
Density 1.0960g/mL
Quantity 5g
Melting Point 7.0°C
Boiling Point 120.0°C to 126.0°C (15.0 mmHg)
Color Brown-Yellow to Yellow
Refractive Index 1.5830 to 1.5850
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5CH=CHCH2Cl
Beilstein 05, II, 372
Fieser 13,299
Solubility Information Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, n,n-dmf,, acetone, formic acid, benzene, ethyl ether
Formula Weight 152.62
Physical Form Liquid
Percent Purity 95% trans
Chemical Name or Material Cinnamyl chloride, 95% trans
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Fatal if inhaled.
May cause an allergic skin reaction.

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Danger

EINECSNumber : 220-246-4