Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cinnamylidenemalonic acid, 98+%

Click to view available options
Quantity:
10g
50g
250g
4472-92-8
C12H10O4
218.208
MFCD00014345
BLPUXJIIRIWMSQ-QPJJXVBHSA-N
cinnamylidenemalonic acid,cinnamalmalonic acid,malonic acid, cinnamylidene,2-e-3-phenylprop-2-enylidene propanedioic acid,propanedioic acid, 3-phenyl-2-propenylidene,2-3-phenylallylidene malonic acid,2-3-phenyl-2-propenylidene malonic acid,2-2e-3-phenylprop-2-en-1-ylidene propanedioic acid,3-phenylprop-2-en-1-ylidene malonic acid,cinnamylidenmalonsaure
822797
2-[(E)-3-phenylprop-2-enylidene]propanedioic acid
C1=CC=C(C=C1)C=CC=C(C(=O)O)C(=O)O
4472-92-8
C12H10O4
218.208
MFCD00014345
BLPUXJIIRIWMSQ-QPJJXVBHSA-N
cinnamylidenemalonic acid,cinnamalmalonic acid,malonic acid, cinnamylidene,2-e-3-phenylprop-2-enylidene propanedioic acid,propanedioic acid, 3-phenyl-2-propenylidene,2-3-phenylallylidene malonic acid,2-3-phenyl-2-propenylidene malonic acid,2-2e-3-phenylprop-2-en-1-ylidene propanedioic acid,3-phenylprop-2-en-1-ylidene malonic acid,cinnamylidenmalonsaure
822797
2-[(E)-3-phenylprop-2-enylidene]propanedioic acid
C1=CC=C(C=C1)C=CC=C(C(=O)O)C(=O)O
Melting Point ~208°C (decomposition)
Quantity 50g
Beilstein 1962423
Formula Weight 218.21
Percent Purity ≥98%
Chemical Name or Material Cinnamylidenemalonic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning