Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CINP Monoclonal Antibody (OTI4E9), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF812210


Description
CINP Monoclonal antibody specifically detects CINP in Human samples. It is validated for Western BlotSpecifications
CINP | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y2Y4 | |
Mouse | |
Protein A/G | |
RUO | |
27033 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI4E9 | |
Unconjugated | |
ZBTB32 | |
1810047K05Rik; 2810452K22Rik; CDK2-interacting protein; Cinp; cyclin dependent kinase 2 interacting protein; cyclin-dependent kinase 2 interacting protein; cyclin-dependent kinase 2-interacting protein; MGC849; RGD1563395 | |
Human recombinant protein fragment corresponding to amino acids 1-300 of human ZBTB32 (NP_055198) produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction