Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CINP Mouse anti-Human, Clone: OTI4E9, liquid, TrueMAB™

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA812210

Encompass_Preferred

Catalog No. 50-168-4454


Add to cart

Description

Description

The function of this gene has not been reported. This gene contains a trinucleotide (ctt) repeat length polymorphism; alleles with either one or two repeat units have been identified.
Specifications

Specifications

CINP
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9BW66
cyclin dependent kinase 2 interacting protein
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51550
Western Blot
OTI4E9
Unconjugated
CINP
Liquid
CINP
Full length human recombit protein of human CINP produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit