Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CIPAR-1 Rabbit anti-Human, HRP, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297050H
Description
CIPAR-1 Polyclonal specifically detects CIPAR-1 in Human samples. It is validated for ELISA.Specifications
CIPAR-1 | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
25849 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
HRP | |
Castration-induced prostatic apoptosis-related protein 1, CIPAR-1, Prostatic androgen-repressed message 1 protein, Protein PARM-1 [Precursor] | |
Produced in rabbits immunized with purified, recombinant CIPAR-1 (AAH13294.1; Met1-Ser258). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title