Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ciprofloxacin Hydrochloride, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
10 kg
25 kg
Packaging:
Fiber Drum
Poly Pail
86393-32-0
C17H21ClFN3O4
385.82
ARPUHYJMCVWYCZ-UHFFFAOYSA-N
hydrogen 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrate chloride
[H+].O.[Cl-].OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O
86393-32-0
C17H21ClFN3O4
385.82
ARPUHYJMCVWYCZ-UHFFFAOYSA-N
hydrogen 1-cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrate chloride
[H+].O.[Cl-].OC(=O)C1=CN(C2CC2)C2=CC(N3CCNCC3)=C(F)C=C2C1=O

C2213, 86393-32-0, C17H18FN3O3•HCl•H2O

Spectrum™ Ciprofloxacin Hydrochloride, USP is used as an antibiotic and in the treatment of bacterial infections. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
Ignition Residue 0.1%
Percent Purity 98-102%
Packaging Poly Pail
Quantity 25 kg