Alfa Aesar™ cis-1,2-Dimethylcyclohexane, 99+%

Manufacturer: Alfa Aesar™ 3146718

Quantity 50g

 View more versions of this product

Catalog No. AA3146718

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2207-01-4
    Formula Weight 112.22
    MDL Number MFCD00063621
    UN Number UN2263
    Chemical Name or Material cis-1,2-Dimethylcyclohexane

    Specifications

    Formula Weight 112.22
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00063621
    CAS 2207-01-4
    UN Number UN2263
    Refractive Index 1.4360
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: DIMETHYLCYCLOHEXANES
    Density 0.796g/mL
    Physical Form Liquid
    Health Hazard 1 Danger
    Boiling Point 129° to 130°C
    Quantity 50g
    Flash Point 12° (54°F)
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H225
    Highly flammable liquid and vapor.
    Chemical Name or Material cis-1,2-Dimethylcyclohexane
    Assay ≥99%

    GHS H Statement
    H225
    Highly flammable liquid and vapor.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit