Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,4-Diacetoxy-2-butene, 95%, ACROS Organics™

Click to view available options
Quantity:
25mL
Packaging:
Glass bottle
25260-60-0
C8H12O4
172.18
MFCD00059339
VZUAUHWZIKOMFC-ARJAWSKDSA-N
cis-1,4-diacetoxy-2-butene,z-but-2-ene-1,4-diyl diacetate,2z-but-2-ene-1,4-diyl diacetate,cis-2-butene-1,4-diol diacetate,2z-4-acetyloxybut-2-enyl acetate,2z-4-acetyloxy but-2-en-1-yl acetate,cis-1,4-diacetoxy-2-buten,z-1,4-diacetoxy-2-butene,cis-2-buten-1,4-diol diacetate,z-but-2-ene-1,4-diyldiacetate
643799
[(Z)-4-acetyloxybut-2-enyl] acetate
CC(=O)OCC=CCOC(=O)C
25260-60-0
C8H12O4
172.18
MFCD00059339
VZUAUHWZIKOMFC-ARJAWSKDSA-N
cis-1,4-diacetoxy-2-butene,z-but-2-ene-1,4-diyl diacetate,2z-but-2-ene-1,4-diyl diacetate,cis-2-butene-1,4-diol diacetate,2z-4-acetyloxybut-2-enyl acetate,2z-4-acetyloxy but-2-en-1-yl acetate,cis-1,4-diacetoxy-2-buten,z-1,4-diacetoxy-2-butene,cis-2-buten-1,4-diol diacetate,z-but-2-ene-1,4-diyldiacetate
643799
[(Z)-4-acetyloxybut-2-enyl] acetate
CC(=O)OCC=CCOC(=O)C
Flash Point >113°C
Packaging Glass bottle
Specific Gravity 1.08
Color Colorless
Boiling Point 222.0°C to 223.0°C
Density 1.0800g/mL
Infrared Spectrum Authentic
Quantity 25mL
Assay Percent Range 94% min. (GC)
Formula Weight 172.18
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material cis-1, 4-Diacetoxy-2-butene
Show More Show Less

GHS H Statement
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
Take off contaminated clothing and wash before reuse.

GHS Signal Word: Warning