Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,4-Dichloro-2-butene, 95%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
Packaging:
Glass bottle
1476-11-5
C4H6Cl2
125
FQDIANVAWVHZIR-UPHRSURJSA-N
cis-1,4-dichloro-2-butene,z-1,4-dichlorobut-2-ene,1,4-dichloro-cis-2-butene,2-butene, 1,4-dichloro-, z,cis-1,4-dichlorobut-2-ene,unii-bqs2752i90,ccris 2651,cis 1,4-dichlorobutene,2-butene, 1,4-dichloro-, cis,cis-2,3-dichlorobut-2-ene
6432115
(Z)-1,4-dichlorobut-2-ene
C(C=CCCl)Cl
1476-11-5
C4H6Cl2
125
FQDIANVAWVHZIR-UPHRSURJSA-N
cis-1,4-dichloro-2-butene,z-1,4-dichlorobut-2-ene,1,4-dichloro-cis-2-butene,2-butene, 1,4-dichloro-, z,cis-1,4-dichlorobut-2-ene,unii-bqs2752i90,ccris 2651,cis 1,4-dichlorobutene,2-butene, 1,4-dichloro-, cis,cis-2,3-dichlorobut-2-ene
6432115
(Z)-1,4-dichlorobut-2-ene
C(C=CCCl)Cl
Melting Point -48.0°C
Refractive Index 1.4884 to 1.4914
Flash Point 55°C
Density 1.1800g/mL
Specific Gravity 1.18
Quantity 25mL
Packaging Glass bottle
Boiling Point 152.0°C (758.0 mmHg)
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Linear Formula ClCH2CH=CHCH2Cl
Beilstein ,III,743
Fieser 10,134; 01,519; 02,229; 03,167; 04,268; 05,357; 06,304; 10,221; 13,152; 17,270
Formula Weight 125
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material cis-1, 4-Dichloro-2-butene
Show More Show Less

GHS Signal Word: Danger

EINECSNumber : 216-021-5