Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,5-Dimethylbicyclo[3.3.0]octane-3,7-dione, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
21170-10-5
C10H14O2
166.22
MFCD00003772
KGZWVKASYOPUET-UHFFFAOYSA-N
cis-1,5-dimethylbicyclo 3.3.0 octane-3,7-dione,3a,6a-dimethylperhydropentalene-2,5-dione,cis-3a,6a-dimethyltetrahydropentalene-2,5 1h,3h-dione,cis-tetrahydro-3a,6a-dimethylpentalene-2,5 1h,3h-dione,3a,6a-dimethyltetrahydro-2,5 1h,3h-pentalenedione,3a,6a-dimethyl-tetrahydropentalene-2,5-dione,3ar,6ar-3a,6a-dimethyl-tetrahydropentalene-2,5-dione,zlchem 111,acmc-20alfk,pubchem6298
89460
3a,6a-dimethyl-1,3,4,6-tetrahydropentalene-2,5-dione
CC12CC(=O)CC1(CC(=O)C2)C
21170-10-5
C10H14O2
166.22
MFCD00003772
KGZWVKASYOPUET-UHFFFAOYSA-N
cis-1,5-dimethylbicyclo 3.3.0 octane-3,7-dione,3a,6a-dimethylperhydropentalene-2,5-dione,cis-3a,6a-dimethyltetrahydropentalene-2,5 1h,3h-dione,cis-tetrahydro-3a,6a-dimethylpentalene-2,5 1h,3h-dione,3a,6a-dimethyltetrahydro-2,5 1h,3h-pentalenedione,3a,6a-dimethyl-tetrahydropentalene-2,5-dione,3ar,6ar-3a,6a-dimethyl-tetrahydropentalene-2,5-dione,zlchem 111,acmc-20alfk,pubchem6298
89460
3a,6a-dimethyl-1,3,4,6-tetrahydropentalene-2,5-dione
CC12CC(=O)CC1(CC(=O)C2)C
Quantity 1g
Melting Point 228.0°C to 230.0°C
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Formula Weight 166.22
Physical Form Powder
Percent Purity 97%
Chemical Name or Material cis-1,5-Dimethylbicyclo[3.3.0]octane-3,7-dione, 98+%

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 244-255-8