Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-2-Pentene, ACROS Organics™

Click to view available options
Quantity:
1g
5g
10g
Packaging:
Glass bottle
627-20-3
C5H10
70.13
MFCD00063978
QMMOXUPEWRXHJS-HYXAFXHYSA-N
cis-2-pentene,z-2-pentene,2-pentene, z,cis-pentene,z-pent-2-ene,unii-54ur3xz4fc,cis-pentene-2,54ur3xz4fc,2-pentene,2-pentene, 2z
5326160
(Z)-pent-2-ene
CCC=CC
627-20-3
C5H10
70.13
MFCD00063978
QMMOXUPEWRXHJS-HYXAFXHYSA-N
cis-2-pentene,z-2-pentene,2-pentene, z,cis-pentene,z-pent-2-ene,unii-54ur3xz4fc,cis-pentene-2,54ur3xz4fc,2-pentene,2-pentene, 2z
5326160
(Z)-pent-2-ene
CCC=CC
Flash Point −27°C
Color Colorless
Packaging Glass bottle
Infrared Spectrum Authentic
Boiling Point 37.0°C
Quantity 1g
Refractive Index 1.3810 to 1.3830 (20°C, 589nm)
Specific Gravity 0.65
Density 0.6500g/mL
Melting Point -151.0°C
Linear Formula CH3CHCHCH2CH3
Merck Index 15, 7235
Solubility Information Solubility in water: insoluble.
Formula Weight 70.13
Physical Form Liquid
Percent Purity ≥95.0%
Chemical Name or Material cis-2-Pentene
Show More Show Less

GHS H Statement:
May be fatal if swallowed and enters airways.
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-988-7