Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-2-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
63216-49-9
C12H21NO4
243.3
MFCD01863244
QJEQJDJFJWWURK-BDAKNGLRSA-N
1r,2s-boc-achc,boc-1,2-cis-achc-oh,boc-cis-2-aminocyclohexanecarboxylic acid,cis-2-tert-butoxycarbonyl amino cyclohexanecarboxylic acid,1r,2s-boc-2-aminocyclohexane carboxylic acid,n-boc-+/--cis-2-amino-cyclohexane-carboxylic acid,cis-2-tert-butoxycarbonylamino-cyclohexanecarboxylic acid,1r,2s-2-tert-butoxycarbonyl amino cyclohexane-1-carboxylic acid,cis-1-t-butyloxycarbonylamino-cyclohexyl-2-carboxylic acid,1r,2s-2-tert-butoxy carbonyl amino cyclohexane-1-carboxylic acid
1512487
(1R,2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
CC(C)(C)OC(=O)NC1CCCCC1C(=O)O
63216-49-9
C12H21NO4
243.3
MFCD01863244
QJEQJDJFJWWURK-BDAKNGLRSA-N
1r,2s-boc-achc,boc-1,2-cis-achc-oh,boc-cis-2-aminocyclohexanecarboxylic acid,cis-2-tert-butoxycarbonyl amino cyclohexanecarboxylic acid,1r,2s-boc-2-aminocyclohexane carboxylic acid,n-boc-+/--cis-2-amino-cyclohexane-carboxylic acid,cis-2-tert-butoxycarbonylamino-cyclohexanecarboxylic acid,1r,2s-2-tert-butoxycarbonyl amino cyclohexane-1-carboxylic acid,cis-1-t-butyloxycarbonylamino-cyclohexyl-2-carboxylic acid,1r,2s-2-tert-butoxy carbonyl amino cyclohexane-1-carboxylic acid
1512487
(1R,2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
CC(C)(C)OC(=O)NC1CCCCC1C(=O)O
Flash Point 194°C
Quantity 1g
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 127°C to 131°C
Assay Percent Range 98%
Linear Formula HOOCC6H10NHCOOC(CH3)3
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol,chloroform
Formula Weight 243.3
Physical Form Amorphous Powder
Percent Purity ≥97.5%
Chemical Name or Material cis-2-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.