Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ cis-3-(Benzyloxycarbonylamino)cyclohexanemethanol, 97%

Click to view available options
Quantity:
250mg
1g
1931962-56-9
C15H21NO3
MFCD09787485
Benzyl cis-3-hydroxymethylcyclohexylcarbamate; cis-3-(Benzyloxycarbonylamino)-1-hydroxymethylcyclohexane
1931962-56-9
C15H21NO3
MFCD09787485
Benzyl cis-3-hydroxymethylcyclohexylcarbamate; cis-3-(Benzyloxycarbonylamino)-1-hydroxymethylcyclohexane
Quantity 250mg
Formula Weight 263.33
Percent Purity 97%
Chemical Name or Material cis-3-(Benzyloxycarbonylamino)cyclohexanemethanol

TSCA : No

Recommended Storage : Ambient temperatures