Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-3-Chloroacrylic acid, 98%, Acros Organics

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1609-93-4
C3H3ClO2
106.505
MFCD00004368
MHMUCYJKZUZMNJ-UPHRSURJSA-N
cis-3-chloroacrylic acid,z-3-chloroacrylic acid,cis-beta-chloroacrylic acid,2-propenoic acid, 3-chloro-, z,cis-3-chloropropenoic acid,2z-3-chloroacrylic acid,2-propenoic acid, 3-chloro-, 2z,cis-.beta.-chloroacrylic acid,z-3-chloroprop-2-enoic acid,acrylic acid, 3-chloro-, z-8ci
643794
CHEBI:27397
(Z)-3-chloroprop-2-enoic acid
C(=CCl)C(=O)O
1609-93-4
C3H3ClO2
106.505
MFCD00004368
MHMUCYJKZUZMNJ-UPHRSURJSA-N
cis-3-chloroacrylic acid,z-3-chloroacrylic acid,cis-beta-chloroacrylic acid,2-propenoic acid, 3-chloro-, z,cis-3-chloropropenoic acid,2z-3-chloroacrylic acid,2-propenoic acid, 3-chloro-, 2z,cis-.beta.-chloroacrylic acid,z-3-chloroprop-2-enoic acid,acrylic acid, 3-chloro-, z-8ci
643794
CHEBI:27397
(Z)-3-chloroprop-2-enoic acid
C(=CCl)C(=O)O
Infrared Spectrum Authentic
Color Beige
Quantity 5g
Packaging Glass bottle
Melting Point 60°C to 64°C
Assay Percent Range 98%
Linear Formula ClCH=CHCO2H
Beilstein 02, 400
Formula Weight 106.51
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material cis-3-Chloroacrylic acid, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 216-545-4