Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-4-Amino-1-cyclohexanecarboxylic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
3685-23-2
C7H13NO2
143.19
MFCD00191730
DRNGLYHKYPNTEA-UHFFFAOYSA-N
4-aminocyclohexanecarboxylic acid,trans-4-aminocyclohexanecarboxylic acid,cis-4-aminocyclohexanecarboxylic acid,cis-4-aminocyclohexane carboxylic acid,cis-4-amino-1-cyclohexanecarboxylic acid,trans-4-amino-cyclohexanecarboxylic acid,cis-4-aminocyclohexanecarboxylicacid,cyclohexanecarboxylic acid, 4-amino-, trans,trans-4-aminocyclohexane carboxylic acid,1s,4s-4-aminocyclohexanecarboxylic acid
171560
4-aminocyclohexane-1-carboxylic acid
C1CC(CCC1C(=O)O)N
3685-23-2
C7H13NO2
143.19
MFCD00191730
DRNGLYHKYPNTEA-UHFFFAOYSA-N
4-aminocyclohexanecarboxylic acid,trans-4-aminocyclohexanecarboxylic acid,cis-4-aminocyclohexanecarboxylic acid,cis-4-aminocyclohexane carboxylic acid,cis-4-amino-1-cyclohexanecarboxylic acid,trans-4-amino-cyclohexanecarboxylic acid,cis-4-aminocyclohexanecarboxylicacid,cyclohexanecarboxylic acid, 4-amino-, trans,trans-4-aminocyclohexane carboxylic acid,1s,4s-4-aminocyclohexanecarboxylic acid
171560
4-aminocyclohexane-1-carboxylic acid
C1CC(CCC1C(=O)O)N
Flash Point >300°C
Color White
Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
Melting Point 278°C to 283°C
Assay Percent Range 97%
Beilstein 14, IV, 981
Solubility Information Solubility in water: soluble
Formula Weight 143.19
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material cis-4-Amino-1-cyclohexanecarboxylic acid, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.