Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-4-Hexen-1-ol, 97%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
928-91-6
C6H12O
100.16
VTIODUHBZHNXFP-IHWYPQMZSA-N
cis-4-hexen-1-ol,z-hex-4-en-1-ol,z-4-hexen-1-ol,4z-hex-4-en-1-ol,4-hexen-1-ol, z,unii-780wma6j84,e-4-hexenol,cis-1-hydroxy-4-hexene,cis-4-hexene-1-ol,4z-4-hexen-1-ol #
5365589
(Z)-hex-4-en-1-ol
CC=CCCCO
928-91-6
C6H12O
100.16
VTIODUHBZHNXFP-IHWYPQMZSA-N
cis-4-hexen-1-ol,z-hex-4-en-1-ol,z-4-hexen-1-ol,4z-hex-4-en-1-ol,4-hexen-1-ol, z,unii-780wma6j84,e-4-hexenol,cis-1-hydroxy-4-hexene,cis-4-hexene-1-ol,4z-4-hexen-1-ol #
5365589
(Z)-hex-4-en-1-ol
CC=CCCCO
Infrared Spectrum Authentic
Density 0.8570g/mL
Boiling Point 70°C (28.0mmHg)
Flash Point 65°C
Specific Gravity 0.857
Quantity 25mL
Color Colorless
Packaging Glass bottle
Assay Percent Range 96% min. (GC)
Formula Weight 100.16
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material cis-4-Hexen-1-ol
Show More Show Less

GHS H Statement:

GHS P Statement:
Do not get in eyes, on skin, or on clothing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 213-187-