Alfa Aesar™ cis-4-Hydroxy-D-proline, 98+%

Manufacturer: Alfa Aesar™ H27633MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27633MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2584-71-6
    Molecular Formula C5H9NO3
    Formula Weight 131.13
    MDL Number MFCD00005252
    Synonym (2R,4R)-(+)-4-Hydroxypyrrolidine-2-carboxylic acid; H-D-cis-Hyp-OH
    Chemical Name or Material cis-4-Hydroxy-D-proline

    Specifications

    Formula Weight 131.13
    Melting Point 243°C
    Molecular Formula C5H9NO3
    MDL Number MFCD00005252
    CAS 2584-71-6
    Synonym (2R,4R)-(+)-4-Hydroxypyrrolidine-2-carboxylic acid; H-D-cis-Hyp-OH
    Quantity 250mg
    Chemical Name or Material cis-4-Hydroxy-D-proline
    Assay ≥98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit