Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-4-Hydroxy-L-proline, 99%, ACROS Organics™

Click to view available options
Quantity:
50mg
250mg
1g
Packaging:
Glass bottle
618-27-9
C5H9NO3
131.131
MFCD00064319
PMMYEEVYMWASQN-IMJSIDKUSA-N
cis-4-hydroxy-l-proline,2s,4s-4-hydroxypyrrolidine-2-carboxylic acid,l-hydroxyproline,l-allohydroxyproline,allo-4-hydroxy-l-proline,cis-4-hydroxyproline,4s-4-hydroxy-l-proline,l-cis-4-hydroxyproline,l-allo-hydroxyproline,l-proline, 4-hydroxy-, 4s
440015
CHEBI:28397
(2S,4S)-4-hydroxypyrrolidine-2-carboxylic acid
C1C(CNC1C(=O)O)O
618-27-9
C5H9NO3
131.131
MFCD00064319
PMMYEEVYMWASQN-IMJSIDKUSA-N
cis-4-hydroxy-l-proline,2s,4s-4-hydroxypyrrolidine-2-carboxylic acid,l-hydroxyproline,l-allohydroxyproline,allo-4-hydroxy-l-proline,cis-4-hydroxyproline,4s-4-hydroxy-l-proline,l-cis-4-hydroxyproline,l-allo-hydroxyproline,l-proline, 4-hydroxy-, 4s
440015
CHEBI:28397
(2S,4S)-4-hydroxypyrrolidine-2-carboxylic acid
C1C(CNC1C(=O)O)O
Specific Rotation Condition −59.00 (20.00°C c=2,H2O)
Specific Rotation -58° to -60° (20°C, 589nm) (c=2, H2O)
Quantity 250mg
Infrared Spectrum Authentic
Packaging Glass bottle
Color White to Beige
Assay Percent Range 99%
Beilstein 22, I, 546
Merck Index 15, 4878
Formula Weight 131.13
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material cis-4-Hydroxy-L-proline, 99%
Show More Show Less

EINECSNumber : 210-542-1

TSCA : TSCA