Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ cis-4-Hydroxycyclohexylamine hydrochloride, 95%

Click to view available options
Quantity:
250mg
1g
56239-26-0
C6H14ClNO
151.634
MFCD07366531
RKTQEVMZBCBOSB-UHFFFAOYSA-N
trans-4-aminocyclohexanol hydrochloride,cis-4-aminocyclohexanol hydrochloride,4-aminocyclohexanol hydrochloride,cis-4-aminocyclohexanol hcl,4-aminocyclohexan-1-ol hydrochloride,cis-4-amino-cyclohexanol hydrochloride,4-amino-cyclohexanol hydrochloride,1r,4r-4-aminocyclohexan-1-ol hydrochloride,cyclohexanol, 4-amino-, hydrochloride,trans-4-aminocyclohexanol hcl
522619
4-aminocyclohexan-1-ol;hydrochloride
C1CC(CCC1N)O.Cl
56239-26-0
C6H14ClNO
151.634
MFCD07366531
RKTQEVMZBCBOSB-UHFFFAOYSA-N
trans-4-aminocyclohexanol hydrochloride,cis-4-aminocyclohexanol hydrochloride,4-aminocyclohexanol hydrochloride,cis-4-aminocyclohexanol hcl,4-aminocyclohexan-1-ol hydrochloride,cis-4-amino-cyclohexanol hydrochloride,4-amino-cyclohexanol hydrochloride,1r,4r-4-aminocyclohexan-1-ol hydrochloride,cyclohexanol, 4-amino-, hydrochloride,trans-4-aminocyclohexanol hcl
522619
4-aminocyclohexan-1-ol;hydrochloride
C1CC(CCC1N)O.Cl
Quantity 250mg
Sensitivity Moisture Sensitive
Melting Point 193°C to 200°C
Beilstein 3909295
Formula Weight 151.63
Physical Form Solid
Percent Purity 95%
Chemical Name or Material cis-4-Hydroxycyclohexylamine hydrochloride

storageNote1 : 2° to 8°C