Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-5-Norbornene-endo-2,3-dicarboxylic acid, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
3853-88-1
C9H10O4
182.175
MFCD00003735
NIDNOXCRFUCAKQ-ZUMNNYEFSA-N
cis-5-norbornene-endo-2,3-dicarboxylic acid
122176685
(2S,3R,4S)-bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid
C1C2C=CC1C(C2C(=O)O)C(=O)O
3853-88-1
C9H10O4
182.175
MFCD00003735
NIDNOXCRFUCAKQ-ZUMNNYEFSA-N
cis-5-norbornene-endo-2,3-dicarboxylic acid
122176685
(2S,3R,4S)-bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid
C1C2C=CC1C(C2C(=O)O)C(=O)O
Packaging Glass bottle
Quantity 1g
Infrared Spectrum Authentic
Color White to Cream
Assay Percent Range 99%
Beilstein 09, II, 576
Formula Weight 182.18
Physical Form Fine Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material cis-5-Norbornene-endo-2, 3-dicarboxylic acid, 99%

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning