Alfa Aesar™ cis-5-Norbornene-endo-2,3-dicarboxylic anhydride, 97%

Manufacturer: Alfa Aesar™ B2495122

Quantity 100g

 View more versions of this product

Catalog No. AAB2495122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 129-64-6
    Molecular Formula C9H8O3
    Formula Weight 164.16
    MDL Number MFCD00151106
    Beilstein 83657
    Merck Index 14,1796
    Synonym Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride; Carbic anhydride
    Chemical Name or Material cis-5-Norbornene-endo-2,3-dicarboxylic anhydride

    Specifications

    Formula Weight 164.16
    Melting Point 164° to 167°C
    Molecular Formula C9H8O3
    MDL Number MFCD00151106
    CAS 129-64-6
    Beilstein 83657
    Merck Index 14,1796
    Synonym Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride; Carbic anhydride
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H334-H318-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes serious eye damage.
    May cause an allergic skin reaction.
    Quantity 100g
    Health Hazard 3 GHS P Statement
    P284a-P261-P280-P305+P351+P338-P310-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number DT5600000
    Chemical Name or Material cis-5-Norbornene-endo-2,3-dicarboxylic anhydride
    Assay 97%

    Description

    Applications
    cis-5-Norbornene-endo-2,3-dicarboxylic anhydride is used as an organic chemical synthesis intermediate.

    Solubility
    It decomposes in water.

    Notes
    Moisture sensitive. Incompatible with oxidizing agents.

    GHS H Statement
    H334-H318-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes serious eye damage.
    May cause an allergic skin reaction.

    GHS P Statement
    P284a-P261-P280-P305+P351+P338-P310-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit