Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ cis-5-Octen-1-ol, 95%, remainder mainly trans-isomer

Click to view available options
Quantity:
5g
25g
64275-73-6
C8H16O
128.215
MFCD00015569
VDHRTASWKDTLER-ONEGZZNKSA-N
oct-5-en-1-ol,e-oct-5-en-1-ol,5-octen-1-ol,5e-oct-5-en-1-ol
6008958
(E)-oct-5-en-1-ol
CCC=CCCCCO
64275-73-6
C8H16O
128.215
MFCD00015569
VDHRTASWKDTLER-ONEGZZNKSA-N
oct-5-en-1-ol,e-oct-5-en-1-ol,5-octen-1-ol,5e-oct-5-en-1-ol
6008958
(E)-oct-5-en-1-ol
CCC=CCCCCO
Flash Point 82°C (179°F)
Refractive Index 1.452
Density 0.85
Quantity 25g
Boiling Point 95°C (25mmHg)
Beilstein 1840670
Formula Weight 128.22
Percent Purity 95%
Chemical Name or Material cis-5-Octen-1-ol
Assay remainder mainly trans-isomer

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 264-764-9

TSCA : Yes

Recommended Storage : Ambient temperatures