Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
cis-ACBD, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent, competitive and selective inhibitor of glutamate uptake. Certain confusion exists over the naming of this compound because of apparent contradictions in the literature. This is the isomer which has the carboxylic acid and the amino groups on the same side of the cyclobutyl ring.
Specifications
Quantity | 10 mg |
Formula Weight | 159.14 |
Percent Purity | >98% |
Chemical Name or Material | cis-ACBD |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction