Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
cis-Aconitic acid, tech. 85%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
cis-Aconitic acid is used as an intermediate in the production of substances pharmaceuticals. It is widely used in chemical research and as reagent. It acts as a cis-isomer of aconitic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
cis-Aconitic acid is used as an intermediate in the production of substances pharmaceuticals. It is widely used in chemical research and as reagent. It acts as a cis-isomer of aconitic acid.
Solubility
Soluble in water, alcohol. Slightly soluble in ether.
cis-Aconitic acid is used as an intermediate in the production of substances pharmaceuticals. It is widely used in chemical research and as reagent. It acts as a cis-isomer of aconitic acid.
Solubility
Soluble in water, alcohol. Slightly soluble in ether.
Specifications
Melting Point | 115°C to 120°C |
Assay Percent Range | tech. 90% |
Quantity | 1 g |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water,alcohol. Slightly soluble in ether. |
IUPAC Name | (Z)-prop-1-ene-1,2,3-tricarboxylic acid |
Formula Weight | 174.11 |
Percent Purity | 90% |
Grade | Technical |
Chemical Name or Material | cis-Aconitic Acid |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement P280g-P305+P351+P338 Wear protective gloves. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. |
EINECSNumber | 209-561-1 |
TSCA | Yes |
Recommended Storage | Keep cold |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction