Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate, 97%, ACROS Organics™

Click to view available options
Quantity:
250mg
500mg
1g
5g
Packaging:
Glass bottle
98014-14-3
C20H16Cl2N4Ru·xH2O
484.35
MFCD00150005
XBOQABVNRXVAKI-UHFFFAOYSA-L
cis-bis 2,2'-bipyridine dichlororuthenium ii hydrate,bis bipyridyl dichlororuthenium hydrate,2,2'-bipyridine-dichlororuthenium-water 2/1/1,cis-dichlorobis 2,2'-bipyridine ruthenium ii,cis-bis 2,2'-bipyridine dichlororuthenium ii monohydrate,cis-bis 2,2-bipyridine dichlororuthenium ii hydrate,cis-bis-2,2'-bipyridine ruthenium ii chloride,hydrate,cis-bis 2,2 inverted exclamation marka-bipyridine dichlororuthenium ii hydrate
16211844
dichlororuthenium;2-pyridin-2-ylpyridine;hydrate
C1=CC=NC(=C1)C2=CC=CC=N2.C1=CC=NC(=C1)C2=CC=CC=N2.O.Cl[Ru]Cl
98014-14-3
C20H16Cl2N4Ru·xH2O
484.35
MFCD00150005
XBOQABVNRXVAKI-UHFFFAOYSA-L
cis-bis 2,2'-bipyridine dichlororuthenium ii hydrate,bis bipyridyl dichlororuthenium hydrate,2,2'-bipyridine-dichlororuthenium-water 2/1/1,cis-dichlorobis 2,2'-bipyridine ruthenium ii,cis-bis 2,2'-bipyridine dichlororuthenium ii monohydrate,cis-bis 2,2-bipyridine dichlororuthenium ii hydrate,cis-bis-2,2'-bipyridine ruthenium ii chloride,hydrate,cis-bis 2,2 inverted exclamation marka-bipyridine dichlororuthenium ii hydrate
16211844
dichlororuthenium;2-pyridin-2-ylpyridine;hydrate
C1=CC=NC(=C1)C2=CC=CC=N2.C1=CC=NC(=C1)C2=CC=CC=N2.O.Cl[Ru]Cl
Packaging Glass bottle
Infrared Spectrum Authentic
Water 11% max. (K.F.)
Quantity 1g
Color Black to Green
Assay Percent Range 96% min. (Based on Carbon)
Solubility Information Solubility in water: soluble
Formula Weight 484.35
Physical Form Powder
Percent Purity 97%
Chemical Name or Material cis-Bis(2, 2'-bipyridine)dichlororuthenium(II) hydrate
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

GHS Signal Word: Warning