cis-Bis-(2,2ft.-bipyridine)dichlororuthenium(II) dihydrate, Ru 19% min., Alfa Aesar™

Manufacturer: Alfa Aesar 1277803

Quantity 1g

 View more versions of this product

Catalog No. AA1277803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15746-57-3
    Physical Form Crystalline
    Assay Percent Range Ru 19% min.
    Molecular Formula C20H16Cl2N4Ru·2H2O
    Formula Weight 520.38
    MDL Number MFCD00012040
    Synonym cis-Dichlorobis(2, 2'-bipyridine)ruthenium(II) dihydrate Ru(bpy)2Cl2·H2O
    Chemical Name or Material cis-Bis-(2, 2 ft.-bipyridine)dichlororuthenium(II) dihydrate

    Specifications

    Formula Weight 520.38
    Quantity 1g
    Color Black
    Physical Form Crystalline
    Assay Percent Range Ru 19% min.
    Molecular Formula C20H16Cl2N4Ru·2H2O
    MDL Number MFCD00012040
    Appearance Black crystalline
    CAS 15746-57-3
    Synonym cis-Dichlorobis(2, 2'-bipyridine)ruthenium(II) dihydrate Ru(bpy)2Cl2·H2O
    Chemical Name or Material cis-Bis-(2, 2 ft.-bipyridine)dichlororuthenium(II) dihydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit