Alfa Aesar™ cis-Dichlorobis(pyridine)platinum(II), 99%

Manufacturer: Alfa Aesar™ B2286501

Quantity 0.25g

 View more versions of this product

Catalog No. AAB2286501

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14872-21-0
    Molecular Formula C10H10Cl2N2Pt
    Formula Weight 424.19
    MDL Number MFCD00058858
    Synonym cis-Bis(pyridine)platinum(II) chloride
    Chemical Name or Material cis-Dichlorobis(pyridine)platinum(II)

    Specifications

    Formula Weight 424.19
    MDL Number MFCD00058858
    Synonym cis-Bis(pyridine)platinum(II) chloride
    Health Hazard 2 GHS H Statement
    H301-H311-H332-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Melting Point 282° to 288°C
    Quantity 0.25g
    Molecular Formula C10H10Cl2N2Pt
    CAS 14872-21-0
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material cis-Dichlorobis(pyridine)platinum(II)
    Assay 99%

    GHS H Statement
    H301-H311-H332-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit